Thallium(I) acetate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Acetic acid thallium (I) salt, Thallous acetate
M-Clarity™ quality level = MQ200
14765-25G
14765-100G
SIAL14765-25GEA
141
EUR
SIAL14765-25G
SIAL14765-100G
Thallium(I) acetate, Sigma-Aldrich®
Thallium (I) acetate
Formula:
C₂H₃O₂Tl MW: 263.43 g/mol Boiling Pt: 1457 °C (1013 hPa) Melting Pt: 110 °C Density: 3.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013045 CAS Number: 563-68-8 EINECS: 209-257-5 UN: 1707 ADR: 6.1,II Merck Index: 12,09392 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...