1,1,2-Trichloroethane, Sigma-Aldrich®
Supplier: Merck
Warning
466212-25ML 466212-1L 466212-250ML
SIAL466212-25MLEA 112 EUR
SIAL466212-25ML SIAL466212-1L SIAL466212-250ML
1,1,2-Trichloroethane, Sigma-Aldrich®
1,1,2-Trichloroethane
Formula: Cl₂HCCH₂Cl
MW: 133.4 g/mol
Boiling Pt: 113.8 °C (1013 hPa)
Melting Pt: –35.5 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000852
CAS Number: 79-00-5
EINECS: 201-166-9
Merck Index: 12,09767