Chloroacetone, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
167479-100G 167479-5G
SIAL167479-100GEA 25.8 EUR
SIAL167479-100G SIAL167479-5G
Chloroacetone, Sigma-Aldrich®
Chloroacetone
Formula: C₃H₅ClO
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS Number: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1,I
Merck Index: 13,02131