Chloroacetaldehyde dimethyl acetal ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-Chloro-2,2-dimethoxyethane
B25049.22 B25049.36
B25049.22EA 29.8 EUR
B25049.22 B25049.36
Chloroacetaldehyde dimethyl acetal ≥97%
Chloroacetaldehyde dimethyl acetal
Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 128…130 °C
Melting Pt: < –73 °C
Density: 1.094
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000948
CAS Number: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3 III