2,2,2-Trichloroethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,2,2-Trichloroethan-1-ol
M-Clarity™ quality level = MQ200
T54801-100G T54801-500G
SIALT54801-100GEA 49.2 EUR
SIALT54801-100G SIALT54801-500G
2,2,2-Trichloroethanol, Sigma-Aldrich®
2,2,2-Trichloroethanol
Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: 17…18 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004677
CAS Number: 115-20-8
EINECS: 204-071-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,09786