2,3-Dichloro-1-propanol, Sigma-Aldrich®
Supplier: Merck
36295-25ML 36295-5ML
SIAL36295-25MLEA 295 EUR
SIAL36295-25ML SIAL36295-5ML
2,3-Dichloro-1-propanol, Sigma-Aldrich®
2,3-Dichloro-1-propanol
Formula: C₃H₆Cl₂O
MW: 128.99 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040446
CAS Number: 616-23-9
EINECS: 210-470-0
UN: 2810
ADR: 6.1,III