8-Chloro-1-octanol 98
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 8-Chloro-1-n-octanol
H28417.AC H28417.AA
H28417.ACEA 140 EUR
H28417.AC H28417.AA
8-Chloro-1-octanol 98
8-Chloro-1-octanol
Purity: 98% CAS No.: 23144-52-7 Molecular Formula: Cl(CH2)8OH Formula Weight: 164.67 Flash Point: 110°(230°F) Refractive Index: 1.458 Boiling Point: 129-130/11mm MDL No.: MFCD00040005
Formula: C₈H₁₇ClO
MW: 164.68 g/mol
Boiling Pt: 129…130 °C (11 mmHg)
Melting Pt: –3 °C
Density: 0.976
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040005
CAS Number: 23144-52-7
EINECS: 245-451-6