1,1,1-Trifluoroacetone 98+%
Supplier: Thermo Fisher Scientific
Danger
139740250 139741000 139745000
ACRO139740250EA 114 EUR
ACRO139740250 ACRO139741000 ACRO139745000
1,1,1-Trifluoroacetone 98+%
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
MW: 112.05 g/mol
Boiling Pt: 22 °C (760 mmHg)
Melting Pt: –78 °C
Density: 1.25 g/cm³
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 2924
ADR: 3 I
Order Now
Specification Test Results
Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥94.0 %