Trifluoroacetic acid ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: TFA
L06374.AC L06374.AE L06374.AP
L06374.ACEA 32.3 EUR
L06374.AC L06374.AE L06374.AP
Trifluoroacetic acid ≥99%
Trifluoroacetic acid
Purity: 99% CAS No.: 76-05-1 Molecular Formula: C2HF3O2 Formula Weight: 114.02 Refractive Index: 1.2840 Odour: Pungent Boiling Point: 72-73° Beilstein Reference: 742035 Merck Reference: 14,9681 UN No.: 2699 MDL No.: MFCD00004169
Formula: CF₃COOH
MW: 114.02 g/mol
Boiling Pt: 72…73 °C
Melting Pt: –15 °C
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM