1-Iodoperfluorbutane ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
A16671.06 A16671.14 A16671.22
A16671.06EA 34.4 EUR
A16671.06 A16671.14 A16671.22
1-Iodoperfluorbutane ≥97%
1-Iodoperfluorbutane
Purity: 97% CAS No.: 423-39-2 Molecular Formula: CF3(CF2)3I Formula Weight: 345.93 Refractive Index: 1.3290 Odour: Pungent Boiling Point: 67° Beilstein Reference: 1777546 UN No.: 3082 MDL No.: MFCD00001062
Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 67 °C
Melting Pt: –89 °C to –87 °C
Density: 2.010
Storage Temperature: Ambient
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8
UN: 3082
ADR: 9,III