Diiodomethane ≥99% stabilised, ReagentPlus®

Supplier: Merck

Synonyms: Methylene iodide

Stabilization: Stabilised with copper

158429-100G 158429-25G 158429-500G
SIAL158429-100GEA 108 EUR
SIAL158429-100G SIAL158429-25G SIAL158429-500G
Diiodomethane ≥99% stabilised, ReagentPlus®
Diiodomethane

Diiodomethane has been used as a probe liquid for evaluation of the polar and dispersive components of the surface energy of the catecholamine coated fiber surfaces.


It may be used for the preparation of cyclopropyl ketones, esters and amides. Diiodomethane may be used as a probe solvent in contact angle measurement to analyze the hydrophilicity of polymer surfaces.

Formula: CH₂I₂
MW: 267.84 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: 6 °C
Density: 3.32537 g/cm³ (20 °C)
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001079
CAS Number: 75-11-6
EINECS: 200-841-5
UN: 2810
ADR: 6.1,III
Merck Index: 12,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37a85a8930c2cbeaf62626d427944a69722cc72&vid=73a1847aa5acf8fa3b0e777fe1bcae9e4f7bde3d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR