Iodomethane stabilised, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Methyl iodide
Stabilization:
Stabilised with silver
67692-100ML
67692-1L
67692-500ML
SIAL67692-100MLEA
79.8
EUR
SIAL67692-100ML
SIAL67692-1L
SIAL67692-500ML
Iodomethane stabilised, Sigma-Aldrich®
Iodomethane
Formula:
CH₃I MW: 141.94 g/mol Boiling Pt: 42.4 °C (1013 hPa) Melting Pt: –66 °C Density: 2.279 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001073 CAS Number: 74-88-4 EINECS: 200-819-5 UN: 2644 ADR: 6.1,I Merck Index: 12,06161 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...