Triethylamine, Sigma-Aldrich®

Supplier: Merck

SIAL90338-10X2MLEA 234 EUR
SIAL90338-10X2ML
Triethylamine, Sigma-Aldrich®
Triethylamine
Formula: (C₂H₅)₃N
MW: 101,19 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0,729 g/cm³ (20 °C)
Flash Pt: –11 °C
MDL Number: MFCD00009051
UN: 1296
ADR: 3,II
Merck Index: 12,09799
https://si.vwr.com/stibo/web/std.lang.all/92/82/729282.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR