Triethylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
90340-2.5L
90340-1L
90340-250ML
90340-4X25ML
SIAL90340-2.5LEA
204
EUR
SIAL90340-2.5L
SIAL90340-1L
SIAL90340-250ML
SIAL90340-4X25ML
Triethylamine, Sigma-Aldrich®
Triethylamine
Formula:
(C₂H₅)₃N Boiling Pt: 90 °C (1013 hPa) Melting Pt: –115 °C Density: 0.729 g/cm³ (20 °C) Flash Pt: –11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3,II Merck Index: 13,09740 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...