Tributylamine, Sigma-Aldrich®
Supplier: Merck
Danger
90780-100ML 90780-500ML
SIAL90780-100MLEA 53 EUR
SIAL90780-100ML SIAL90780-500ML
Tributylamine, Sigma-Aldrich®
Tributylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
Boiling Pt: 216
Melting Pt: –70 °C
Density: 0.7771 g/cm³ (20 °C)
Flash Pt: 63
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS Number: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1 II
Merck Index: 13,09691