Tributylamine, Sigma-Aldrich®
Supplier: Merck
Danger
90781-10ML 90781-50ML
SIAL90781-10MLEA 43.8 EUR
SIAL90781-10ML SIAL90781-50ML
Tributylamine, Sigma-Aldrich®
Tributylamine
Formula: C₁₂H₂₇N
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.7771 g/cm³ (20 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS Number: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1,II
Merck Index: 13,09691