Hexyl isocyanate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

414883-25ML 414883-5ML
SIAL414883-25MLEA 373 EUR
SIAL414883-25ML SIAL414883-5ML
Hexyl isocyanate, Sigma-Aldrich®
Hexyl isocyanate
Formula: C₇H₁₃NO
Boiling Pt: 44 °C (1013 hPa)
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037047
CAS Number: 2525-62-4
EINECS: 219-763-8
UN: 3080
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR