Dodecylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Dodecylamine, Dodecan-1-amine, 1-Aminododecane
M-Clarity™ quality level = MQ200
44172-25G 44172-5G
SIAL44172-25GEA 205 EUR
SIAL44172-25G SIAL44172-5G
Dodecylamine, Sigma-Aldrich®
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
MW: 185.35 g/mol
Boiling Pt: 247
Melting Pt: 25…28 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8 II