Hexylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Hexylamine, 1-Aminohexane
M-Clarity™ quality level = MQ200
219703-100ML 219703-5ML
SIAL219703-100MLEA 37.7 EUR
SIAL219703-100ML SIAL219703-5ML
Hexylamine, Sigma-Aldrich®
Hexylamine
Formula: H₃C(CH₂)₄CH₂NH₂
MW: 101.19 g/mol
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: –19 °C
Density: 0.766 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008240
CAS Number: 111-26-2
EINECS: 203-851-8
UN: 2734
ADR: 8,I