Octadecylamine, Sigma-Aldrich®

Supplier: Merck

SIAL74750-100GEA 298 EUR
SIAL74750-100G SIAL74750-25G
Octadecylamine, Sigma-Aldrich®
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269,51 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 0,8618 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008159
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR