Acetonedimethyl acetal, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane
M-Clarity™ quality level = MQ200
D136808-25ML D136808-2.5L D136808-500ML D136808-18L
SIALD136808-25MLEA 25.9 EUR
SIALD136808-25ML SIALD136808-2.5L SIALD136808-500ML SIALD136808-18L
Acetonedimethyl acetal, Sigma-Aldrich®
Acetone-dimethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.845 g/cm³ (20 °C)
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II