Formaldehyde dimethyl acetal, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Formal, Methylal, Dimethoxymethane

M-Clarity™ quality level = MQ200

D134651-100ML D134651-1L D134651-2.5L
SIALD134651-100MLEA 29.1 EUR
SIALD134651-100ML SIALD134651-1L SIALD134651-2.5L
Formaldehyde dimethyl acetal, Sigma-Aldrich®
Formaldehyde dimethyl acetal
Formula: CH₂(OCH₃)₂
MW: 76.1 g/mol
Boiling Pt: 42 °C (1013 hPa)
Melting Pt: –105 °C
Density: 0.821 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008495
CAS Number: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: 3,II
Merck Index: 12,06093
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dc068e2803e769f0f748f5152cbff2cbfe69fc&vid=eae965f4719d855b0e2585391d097f07b53b9067&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR