1,5-Pentanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane
M-Clarity™ quality level = MQ200
76892-1L 76892-250ML 76892-25L
SIAL76892-1LEA 141 EUR
SIAL76892-1L SIAL76892-250ML SIAL76892-25L
1,5-Pentanediol, Sigma-Aldrich®
1,5-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: –16 °C
Density: 0.9941 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002978
CAS Number: 111-29-5
EINECS: 203-854-4
Merck Index: 12,07256