1-Tetradecanethiol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
n-Tetradecyl mercaptan, 1-Mercaptotetradecane, n-Tetradecanethiol, N-tetradecylmercaptan
87193-25ML
87193-5ML
SIAL87193-25MLEA
211
EUR
SIAL87193-25ML
SIAL87193-5ML
1-Tetradecanethiol, Sigma-Aldrich®
1-Tetradecanethiol
Formula:
C₁₄H₃₀S MW: 230.46 g/mol Boiling Pt: 118 °C (0.5 mmHg) Melting Pt: 7 °C Density: 0.8641 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00022099 CAS Number: 2079-95-0 EINECS: 218-209-2 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...