2-Propanethiol 98%
Supplier: Thermo Fisher Scientific
Danger
131440500 131445000 131440025
ACRO131440500EA 23.9 EUR
ACRO131440500 ACRO131445000 ACRO131440025
2-Propanethiol 98%
2-Propanethiol
Formula: C₃H₈S
Boiling Pt: 57…60 °C (760 mmHg)
Melting Pt: –131 °C
Density: 0.82 g/cm³
Flash Pt: –34 °C
Storage Temperature: Ambient
MDL Number: MFCD00004863
CAS Number: 75-33-2
EINECS: 200-861-4
UN: 2402
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4270 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤15 APHA