n-Docosane ≥99%

Supplier: Thermo Scientific

Synonyms: N-docosane

A18050.14 A18050.22
A18050.14EA 43.4 EUR
A18050.14 A18050.22
n-Docosane ≥99%
n-Docosane

Purity: 99% CAS No.: 629-97-0 Molecular Formula: CH3(CH2)20CH3 Formula Weight: 310.61 Flash Point: 110°(230°F) Refractive Index: 1.4455 Melting Point: 43-46° Boiling Point: 368-369° Beilstein Reference: 1702206 MDL No.: MFCD00009348

Formula: H₃C(CH₂)₂₀CH₃
MW: 310.61 g/mol
Boiling Pt: 368…369 °C
Melting Pt: 43…46 °C
Density: 0.777
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR