n-Heptadecane 99%

Supplier: Thermo Scientific
Danger

Synonyms: N-heptadecane

156240250 156241000 156245000
ACRO156240250EA 47.6 EUR
ACRO156240250 ACRO156241000 ACRO156245000
n-Heptadecane 99%
n-Heptadecane
Formula: H₃C(CH₂)₁₅CH₃
MW: 240.47 g/mol
Boiling Pt: 302 °C (760 mmHg)
Melting Pt: 22…24 °C
Density: 0.778 g/cm³
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00009002
CAS Number: 629-78-7
EINECS: 211-108-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
GC ≥98.5 %
Chlorine (Cl) ≤0.05 %
Iodine (I) ≤0.001 %
Nickel (Ni) ≤5 ppm
Heavy metals (as Pb) ≤2 ppm
Sulfur compounds (as S) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR