n-Hexane ∼95% (n-hexane) for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Danger
326660010 326660025
ACRO326660010EA 57.6 EUR
ACRO326660010 ACRO326660025
n-Hexane ∼95% (n-hexane) for pesticide residue analysis
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.01 %
GC (ECD) Passes test, (NPD) Passes test, (FID) Passes test
Residue after evaporation ≤2 ppm