n-Hexane, dry_extra 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Hexyl hydride, Hexanen, Hexane
326921000 326920010 326920025
326921000.EA 31 EUR
326921000. 326920010. 326920025.
n-Hexane, dry_extra 96+%, AcroSeal®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.002 %
GC ≥96.0 %
Residue after evaporation ≤0.0005 %
Filt.over 0.2 micron PFTE Filtered