n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
364371000 364370010 364370025 364375000
364371000.EA 31.5 EUR
364371000. 364370010. 364370025. 364375000.
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥96.0 %