n-Nonane

Supplier: Roth Carl

ROTH4310.1EA 53.5 EUR
ROTH4310.1
n-Nonane
n-Nonane
Formula: H₃C(CH₂)₇CH₃
MW: 128,26 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –53 °C
Density: 0,718 g/cm³ (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00009574
UN: 1920
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR