3-Phenyl-1-propyne 97% stabilised
Supplier: Thermo Scientific
Warning
Synonyms: 2-Propynylbenzene
376800010 376800050 376800250
ACRO376800010EA 68 EUR
ACRO376800010 ACRO376800050 ACRO376800250
3-Phenyl-1-propyne 97% stabilised
3-Phenyl-1-propyne
Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 75 °C (20 mmHg)
Density: 0.934 g/cm³
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00134431
CAS Number: 10147-11-2
UN: 3295
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5260 to 1.5300 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 100 to 400 ppm BHT