2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 6-Bromo-α,α,α,4-tetrafluoro-o-toluidine
H26206.03 H26206.09
H26206.03EA 32.1 EUR
H26206.03 H26206.09
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline
Purity: 98% CAS No.: 875664-27-0 Molecular Formula: C7H4BrF4N Formula Weight: 258.01 Refractive Index: 1.503
Formula: C₇H₄BrF₄N
MW: 258.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973756
CAS Number: 875664-27-0
EINECS: None
UN: 2810
ADR: 6.1 III