2'-Hydroxy-5'-methylacetophenone

Supplier: Thermo Scientific

Synonyms: 1-(2-Hydroxy-5-methylphenyl)ethanone

BTB10270.1 BTB10270.10 BTB10270.50
MAYBBTB10270.1EA 0 EUR
MAYBBTB10270.1 MAYBBTB10270.10 MAYBBTB10270.50
2'-Hydroxy-5'-methylacetophenone
2'-Hydroxy-5'-methylacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 83…87 °C (0.5 mmHg)
Melting Pt: 53 °C
Density: 1,0797 g/cm³ (53 °C)
MDL Number: MFCD00002380
CAS Number: 1450-72-2
EINECS: 215-915-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR