4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Supplier: Thermo Scientific
SB01186.1
SB01186.10
SB01186.50
MAYBSB01186.1EA
0
EUR
MAYBSB01186.1
MAYBSB01186.10
MAYBSB01186.50
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Formula:
C₁₀H₇F₃O₂ |
MDL Number:
MFCD00000425 CAS Number: 326-06-7 EINECS: 206-307-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...