1-(Trimethylsilyl)-1-propyne, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethyl-1-propynylsilane
M-Clarity™ quality level = MQ100
244481-5G 244481-25G
SIAL244481-5GEA 128 EUR
SIAL244481-5G SIAL244481-25G
1-(Trimethylsilyl)-1-propyne, Sigma-Aldrich®
1-(Trimethylsilyl)-1-propyne
Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 99 °C (1013 hPa)
Density: 0.759 g/cm³ (20 °C)
MDL Number: MFCD00009271
CAS Number: 6224-91-5
EINECS: 228-314-5