3-Chloro-1-phenyl-1-propyne, Sigma-Aldrich®
Supplier: Merck
Synonyms: (3-Chloroprop-1-yn-1-yl)benzene
M-Clarity™ quality level = MQ100
664960-25G 664960-5G
SIAL664960-25GEA 375 EUR
SIAL664960-25G SIAL664960-5G
3-Chloro-1-phenyl-1-propyne, Sigma-Aldrich®
3-Chloro-1-phenyl-1-propyne
Formula: C₉H₇Cl
MW: 150.61 g/mol
MDL Number: MFCD06411085
CAS Number: 3355-31-5