trans,trans-1,4-Diphenyl-1,3-butadiene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (1E,3E)-1,4-Diphenylbuta-1,3-diene
M-Clarity™ quality level = MQ200
D206008-25G D206008-5G
SIALD206008-25GEA 118 EUR
SIALD206008-25G SIALD206008-5G
trans,trans-1,4-Diphenyl-1,3-butadiene, Sigma-Aldrich®
trans,trans-1,4-Diphenyl-1,3-butadiene
Formula: C₁₆H₁₄
MW: 206.29 g/mol
MDL Number: MFCD00004791
CAS Number: 538-81-8