Propagyl bromide 80% ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Bromopropyne
P51001-5KG P51001-125G P51001-50G P51001-625G
SIALP51001-5KGEA 4890 EUR
SIALP51001-5KG SIALP51001-125G SIALP51001-50G SIALP51001-625G
Propagyl bromide 80% ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 90
Melting Pt: -43
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3 II