VWR® Cleanroom Crimping Tools
Supplier: VWR
VWR®
548-1377EA 814 EUR
548-1377 548-1485 548-1376
VWR® Cleanroom Crimping Tools
Crimpers, Cappers and Decappers
Made entirely of stainless steel for maximum durability during dry heat sterilisation or autoclaving.
  • Corrosion and heat resistant, no need for any protective coating on the handle or crimp head
  • Withstands repeated sterilisation for cleanroom use without the risk of damage
  • Adjustable crimping pressure and height