n-Tetradecane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tetradecane
87140-250ML 87140-50ML
SIAL87140-250MLEA 118 EUR
SIAL87140-250ML SIAL87140-50ML
n-Tetradecane, Sigma-Aldrich®
n-Tetradecane
Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Boiling Pt: 253.7
Melting Pt: 6
Density: 0.7628 g/cm³ (20 °C)
Flash Pt: 102 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM