1-Bromo-3,3-dimethylbutanone
Supplier: Thermo Scientific
Warning
Synonyms: 1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone
CD08380.1 CD08380EA CD08380.50
MAYBCD08380.1EA 0 EUR
MAYBCD08380.1 MAYBCD08380EA MAYBCD08380.50
1-Bromo-3,3-dimethylbutanone
1-Bromo-3,3-dimethylbutanone
Formula: C₆H₁₁BrO
MW: 179.06 g/mol
Boiling Pt: 47…49 °C (1 mmHg)
Melting Pt: 76
Density: 1.3508 g/cm³ (0 °C)
Storage Temperature: Freezer
MDL Number: MFCD00000206
CAS Number: 5469-26-1
EINECS: 226-794-0
UN: 2810
ADR: 6.1 II