Isovaleraldehyde 98%
Supplier: Thermo Fisher Scientific
Danger
163971000 163975000 163970025
ACRO163971000EA 22.2 EUR
ACRO163971000 ACRO163975000 ACRO163970025
Isovaleraldehyde 98%
Isovaleraldehyde
Formula: C₅H₁₀O
Boiling Pt: 90 °C (760 mmHg)
Melting Pt: –28 °C
Density: 0.796 g/cm³
Flash Pt: –5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: 3,II
Merck Index: 13,05250
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20°C, 589 nm)
GC ≥97.5 %