1-Chloro-2-methyl-2-propanol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A18978.14
A18978.22
A18978.14EA
65.3
EUR
A18978.14
A18978.22
1-Chloro-2-methyl-2-propanol ≥98%
1-Chloro-2-methyl-2-propanol
Purity: 98%
CAS No.: 558-42-9
Molecular Formula: C4H9ClO
Formula Weight: 108.57
Flash Point: 38°(100°F)
Refractive Index: 1.4400
Boiling Point: 124-128°
Beilstein Reference: 1697054
UN No.: 1987
MDL No.: MFCD00021807
Formula:
C₄H₉ClO Boiling Pt: 124…128 °C Melting Pt: –20 °C Density: 1.058 Flash Pt: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021807 CAS Number: 558-42-9 EINECS: 209-196-4 UN: 1987 ADR: 3,III |