2,2,4,4,6,8,8-Heptamethylnonane 98%

Supplier: Thermo Fisher Scientific
Danger

156250500 156250010 156252500
ACRO156250500EA 68.3 EUR
ACRO156250500 ACRO156250010 ACRO156252500
2,2,4,4,6,8,8-Heptamethylnonane 98%
2,2,4,4,6,8,8-Heptamethylnonane
Formula: C₁₆H₃₄
MW: 226.45 g/mol
Boiling Pt: 240 °C (760 mmHg)
Density: 0.79 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008856
CAS Number: 4390-04-9
EINECS: 224-506-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR