Hand pads, ultra fine, 7448, Scotch-Brite™
Supplier: 3M
07448_U 07448
115-3007EA 26.5 EUR
115-3007 115-1752
Hand pads, ultra fine, 7448, Scotch-Brite™
Wipes Cleaning Kits and Pads
These hand pads can be used in place of steel wool for conditioning metal surfaces and paint prep. Unlike fine steel wool, Scotch-Brite™ pads will not shred during use, rust after use, or leave fine metal splinters.
  • Performs like steel wool grade 00 without shredding, rusting, or splintering
  • Pad conforms to irregular surfaces and gets into corners without losing performance
  • Cleans metal, composites, and other materials without altering the base material
  • Load and heat resistant non woven web provides consistent finishing over the life of the pad
Ideal for cleaning, blending, prepping and finishing on metal, wood, plastics and composites.