Hand pads, pre-cut rolls, Scotch-Brite™
Supplier: 3M
Scotch-Brite™
MMMA07904EA 304 EUR
MMMA07904
Hand pads, pre-cut rolls, Scotch-Brite™
Wipes Cleaning Kits and Pads
Pre perforated, ready-to use Scotch-Brite™ Hand Pads with a new super sharp 3M abrasive for more aggressive cut without increased coarseness. They may be used for sanding awkward areas before applying etch primer, for example in the transportation industry or during other repair processes. The pads are easy to dispense one by one.
  • Longer lasting, more economical
  • Faster, easier scuffing
  • Improved abrasive, ultimate finish