2-Methylheptane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Isoheptane
M-Clarity™ quality level = MQ100
M47949-5G
M47949-25G
SIALM47949-5GEA
140
EUR
SIALM47949-5G
SIALM47949-25G
2-Methylheptane, Sigma-Aldrich®
2-Methylheptane
Formula:
C₈H₁₈ MW: 114.23 g/mol Boiling Pt: 117.2 Melting Pt: –109 °C Density: 0.698 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009518 CAS Number: 592-27-8 EINECS: 209-747-9 UN: 1262 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...