Allyl bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Bromopropene, 3-Propenyl bromide
M-Clarity™ quality level = MQ200
A29585-100G A29585-1KG A29585-3KG A29585-500G A29585-5G
SIALA29585-100GEA 53.6 EUR
SIALA29585-100G SIALA29585-1KG SIALA29585-3KG SIALA29585-500G SIALA29585-5G
Allyl bromide, Sigma-Aldrich®
Allyl bromide
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 70
Melting Pt: –119 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: –1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000244
CAS Number: 106-95-6
EINECS: 203-446-6
UN: 1099
ADR: 3 I
Merck Index: 13,00285