Vinyl acetate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Acetic acid vinyl ester
M-Clarity™ quality level = MQ200
V1503-1L
V1503-2.5L
V1503-25ML
V1503-18L
SIALV1503-1LEA
38.3
EUR
SIALV1503-1L
SIALV1503-2.5L
SIALV1503-25ML
SIALV1503-18L
Vinyl acetate, Sigma-Aldrich®
Vinyl acetate
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 72.3 °C (1013 hPa) Melting Pt: –100 °C Density: 0.932 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008713 CAS Number: 108-05-4 EINECS: 203-545-4 UN: 1301 ADR: 3,II Merck Index: 12,10130 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...